2-Chloro-4-iodopyridin-3-ol 97%

Lieferant: Apollo Scientific

OR968326-1G
APOSOR968326-1GEA 180 EUR
APOSOR968326-1G
2-Chloro-4-iodopyridin-3-ol 97%
2-Chloro-4-iodopyridin-3-ol
Formel: C₅H₃ClINO
Lagertemperatur: Kühlschrank
MDL: MFCD16610456
CAS-Nummer: 1207973-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR