3-(2-Methyl-1,3-thiazol-4-yl)benzonitrile

Lieferant: Apollo Scientific

OR9688-1G
APOSOR9688-1GEA 87 EUR
APOSOR9688-1G
3-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
3-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
Formel: C₁₁H₈N₂S
Molecular Weight: 200,26 g/mol
Schmelzpunkt: 117…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659064
CAS-Nummer: 844891-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR