1-(2-Bromophenyl)-1H-pyrazole 97%

Lieferant: Apollo Scientific
Achtung

OR9690-1G
APOSOR9690-1GEA 126 EUR
APOSOR9690-1G
1-(2-Bromophenyl)-1H-pyrazole 97%
1-(2-Bromophenyl)-1H-pyrazole
Formel: C₉H₇BrN₂
Molecular Weight: 223,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659065
CAS-Nummer: 87488-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR