Imidazo[1,2-a]pyridin-3-carbonsäure 98%

Lieferant: Apollo Scientific
Achtung

OR9691-1G
APOSOR9691-1GEA 21.9 EUR
APOSOR9691-1G
Imidazo[1,2-a]pyridin-3-carbonsäure 98%
Imidazo[1,2-a]pyridin-3-carbonsäure
Formel: C₈H₆N₂O₂
Molecular Weight: 162.1454 g/mol
Schmelzpunkt: 195.5…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01720458
CAS-Nummer: 6200-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR