4-(4-Formylbenzyl)piperazine, N1-BOC protected 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzaldehyde, tert-Butyl 4-(4-formylbenzyl)piperazine-1-carboxylate
OR9694-250MG
APOSOR9694-250MGEA
62.5
EUR
APOSOR9694-250MG
4-(4-Formylbenzyl)piperazine, N1-BOC protected 95%
4-(4-Formylbenzyl)piperazine, N1-BOC protected
Formel:
C₁₇H₂₄N₂O₃ Schmelzpunkt: 57…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659067 CAS-Nummer: 844891-09-4 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...