4-[4-(Hydroxymethyl)benzyl]piperazine, N1-BOC protected 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: tert-Butyl 4-(4-(hydroxymethyl)benzyl)piperazine-1-carboxylate, tert-Butyl 4-[4-(hydroxymethyl)benzyl]piperazine-1-carboxylate

OR9695-1G
APOSOR9695-1GEA 551 EUR
APOSOR9695-1G
4-[4-(Hydroxymethyl)benzyl]piperazine, N1-BOC protected 95%
4-[4-(Hydroxymethyl)benzyl]piperazine, N1-BOC protected
Formel: C₁₇H₂₆N₂O₃
Molecular Weight: 306,4 g/mol
Schmelzpunkt: 52…53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06658975
CAS-Nummer: 622381-67-3
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR