4-[4-(Methoxycarbonyl)benzyl]piperazine, N1-BOC protected

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-[4(methoxycarbonyl)benzyl]piperazine-1-carboxylate

OR9698-1G
APOSOR9698-1GEA 125 EUR
APOSOR9698-1G
4-[4-(Methoxycarbonyl)benzyl]piperazine, N1-BOC protected
4-[4-(Methoxycarbonyl)benzyl]piperazine, N1-BOC protected
Formel: C₁₈H₂₆N₂O₄
Schmelzpunkt: 72,5…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06658976
CAS-Nummer: 844891-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR