(4-Methylthien-2-yl)methylamine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9699-1G
OR9699-250MG
APOSOR9699-1GEA
315
EUR
APOSOR9699-1G
APOSOR9699-250MG
(4-Methylthien-2-yl)methylamine 97%
(4-Methylthien-2-yl)methylamine
Formel:
C₆H₉NS Siedepunkt: 42…46 °C/0,1mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD06657973 CAS-Nummer: 104163-39-5 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...