1-[4-Methyl-2-(phenylamino)-1,3-thiazol-5-yl]ethan-1-one 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Methyl-2-(phenylamino)thiazol-5-yl)ethanone, 5-Acetyl-2-anilino-4-methyl-1,3-thiazole, 5-Acetyl-4-methyl-2-(phenylamino)-1,3-thiazole
OR9700-1G
APOSOR9700-1GEA
58
EUR
APOSOR9700-1G
1-[4-Methyl-2-(phenylamino)-1,3-thiazol-5-yl]ethan-1-one 98%
1-[4-Methyl-2-(phenylamino)-1,3-thiazol-5-yl]ethan-1-one
Formel:
C₁₂H₁₂N₂OS Molecular Weight: 232,301 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00129318 CAS-Nummer: 31609-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...