4-Ethylphenylhydrazin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR9704-25G
APOSOR9704-25GEA 157 EUR
APOSOR9704-25G
4-Ethylphenylhydrazin Hydrochlorid
4-Ethylphenylhydrazin Hydrochlorid
Formel: C₈H₁₃ClN₂
Schmelzpunkt: 232…234 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01861267
CAS-Nummer: 53661-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR