3-(1H-Imidazol-1-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

OR9721-5G
APOSOR9721-5GEA 264 EUR
APOSOR9721-5G
3-(1H-Imidazol-1-yl)benzoic acid 97%
3-(1H-Imidazol-1-yl)benzoic acid
Formel: C₁₀H₈N₂O₂
Schmelzpunkt: 194…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659077
CAS-Nummer: 108035-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR