3-(1H-Pyrazol-1-yl)benzoesäure 98%

Lieferant: Apollo Scientific
Achtung

OR9722-1G
APOSOR9722-1GEA 101 EUR
APOSOR9722-1G
3-(1H-Pyrazol-1-yl)benzoesäure 98%
3-(1H-Pyrazol-1-yl)benzoic acid
Formel: C₁₀H₈N₂O₂
Schmelzpunkt: 154…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05864484
CAS-Nummer: 264264-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR