5-Bromo-3-(bromoacetyl)benzo[b]thiophene 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-1-(5-bromo-1-benzothiophen-3-yl)ethan-1-one, 5-Bromo-3-(bromoacetyl)-1-benzothiophene
OR9723-1G
APOSOR9723-1GEA
187
EUR
APOSOR9723-1G
5-Bromo-3-(bromoacetyl)benzo[b]thiophene 97%
5-Bromo-3-(bromoacetyl)benzo[b]thiophene
Formel:
C₁₀H₆Br₂OS Molecular Weight: 334,027 g/mol Schmelzpunkt: 156…157 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06797474 CAS-Nummer: 850375-12-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...