2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Carboxyphenyl)-2,5-dimethyl-1H-pyrrole

OR9731-1G OR9731-250MG
APOSOR9731-1GEA 143 EUR
APOSOR9731-1G APOSOR9731-250MG
2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid 97%
2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid
Formel: C₁₃H₁₃NO₂
Schmelzpunkt: 125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01125264
CAS-Nummer: 92028-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR