2-Brom-6-nitrophenol 98%

Lieferant: Apollo Scientific
Achtung

OR9738-100G OR9738-25G OR9738-5G
APOSOR9738-100GEA 85.7 EUR
APOSOR9738-100G APOSOR9738-25G APOSOR9738-5G
2-Brom-6-nitrophenol 98%
2-Brom-6-nitrophenol
Formel: C₆H₄BrNO₃
Molecular Weight: 218.01 g/mol
Schmelzpunkt: 66…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683216
CAS-Nummer: 13073-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR