5-Iodisatin 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR9765-100G
OR9765-25G
OR9765-5G
APOSOR9765-100GEA
270
EUR
APOSOR9765-100G
APOSOR9765-25G
APOSOR9765-5G
5-Iodisatin 97%
5-Iodisatin
Formel:
C₈H₄INO₂ Schmelzpunkt: 276…280 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016899 CAS-Nummer: 20780-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...