4-[4-(Hydroxymethyl)pyridin-2-yl]morpholine 97%
Lieferant: Apollo Scientific
OR9782-250MG
APOSOR9782-250MGEA
0
EUR
APOSOR9782-250MG
4-[4-(Hydroxymethyl)pyridin-2-yl]morpholine 97%
4-[4-(Hydroxymethyl)pyridin-2-yl]morpholine
Formel:
C₁₀H₁₄N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08271915 CAS-Nummer: 556109-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...