Ethyl 1-methyl-3-phenyl-1H-pyrazole-5-carboxylate 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[5-(Ethoxycarbonyl)-1-methyl-1H-pyrazol-3-yl]benzene, 5-(Ethoxycarbonyl)-1-methyl-3-phenyl-1H-pyrazole
OR9786-1G
APOSOR9786-1GEA
81.2
EUR
APOSOR9786-1G
Ethyl 1-methyl-3-phenyl-1H-pyrazole-5-carboxylate 95+%
Ethyl 1-methyl-3-phenyl-1H-pyrazole-5-carboxylate
Formel:
C₁₃H₁₄N₂O₂ Molecular Weight: 230,262 g/mol Schmelzpunkt: 27 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03933288 CAS-Nummer: 10250-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...