2-Methylquinoline-6-carbaldehyde 97%

Lieferant: Apollo Scientific
Achtung

OR9788-250MG
APOSOR9788-250MGEA 65.4 EUR
APOSOR9788-250MG
2-Methylquinoline-6-carbaldehyde 97%
2-Methylquinoline-6-carbaldehyde
Formel: C₁₁H₉NO
Molecular Weight: 171.20 g/mol
Schmelzpunkt: 96…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08060502
CAS-Nummer: 108166-03-6
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR