3-Isopropoxybenzoic acid

Lieferant: Apollo Scientific
Achtung

OR9794-1G OR9794-5G
APOSOR9794-1GEA 0 EUR
APOSOR9794-1G APOSOR9794-5G
3-Isopropoxybenzoic acid
3-Isopropoxybenzoic acid
Formel: C₁₀H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01922170
CAS-Nummer: 60772-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR