2,5-Di-n-butoxy-4-morpholinobenzenediazonium p-toluenesulphonate

Lieferant: Apollo Scientific

OR98028
APOSOR98028EA 883 EUR
APOSOR98028
2,5-Di-n-butoxy-4-morpholinobenzenediazonium p-toluenesulphonate
2,5-Di-n-butoxy-4-morpholinobenzenediazonium p-toluenesulphonate
Formel: C₂₅H₃₅N₃O₆S
CAS-Nummer: 93672-52-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR