Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)

Lieferant: Apollo Scientific

OR9809-1G OR9809-5G
APOSOR9809-1GEA 72.5 EUR
APOSOR9809-1G APOSOR9809-5G
Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)
Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)
Formel: C₁₄H₂₀O₅
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005945
CAS-Nummer: 14098-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR