4-(2-Phenylacetyl)-1H-pyrrole-2-carboxylic acid 96%

Lieferant: Apollo Scientific
Achtung

OR9816-250MG
APOSOR9816-250MGEA 743 EUR
APOSOR9816-250MG
4-(2-Phenylacetyl)-1H-pyrrole-2-carboxylic acid 96%
4-(2-Phenylacetyl)-1H-pyrrole-2-carboxylic acid
Formel: C₁₃H₁₁NO₃
Molecular Weight: 229,24 g/mol
Schmelzpunkt: 236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02571500
CAS-Nummer: 478249-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR