1-Methyl-3-(3-nitrophenyl)pyrazole

Lieferant: Apollo Scientific

OR9822-1G
APOSOR9822-1GEA 265 EUR
APOSOR9822-1G
1-Methyl-3-(3-nitrophenyl)pyrazole
1-Methyl-3-(3-nitrophenyl)pyrazole
Formel: C₁₀H₉N₃O₂
Molecular Weight: 203,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665958
CAS-Nummer: 886851-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR