2-Butyl-5-(4-nitrophenyl)-1,3,4-oxadiazole

Lieferant: Apollo Scientific

OR9828-1G
APOSOR9828-1GEA 154 EUR
APOSOR9828-1G
2-Butyl-5-(4-nitrophenyl)-1,3,4-oxadiazole
2-Butyl-5-(4-nitrophenyl)-1,3,4-oxadiazole
Formel: C₁₂H₁₃N₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06797384
CAS-Nummer: 100933-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR