6-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR9830-1G
APOSOR9830-1GEA
81.2
EUR
APOSOR9830-1G
6-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
6-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Formel:
C₈H₇ClN₄O Schmelzpunkt: 228…230 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797385 CAS-Nummer: 119448-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...