2-(tert-Butyl)-1H-pyrrolo[2,3-b]pyridine 95+%
Lieferant: Apollo Scientific
Synonyme:
2-(tert-Butyl)-7-azaindole
OR9836-10G
OR9836-1G
OR9836-5G
APOSOR9836-10GEA
575
EUR
APOSOR9836-10G
APOSOR9836-1G
APOSOR9836-5G
2-(tert-Butyl)-1H-pyrrolo[2,3-b]pyridine 95+%
2-(tert-Butyl)-1H-pyrrolo[2,3-b]pyridine
Formel:
C₁₁H₁₄N₂ Molecular Weight: 174.25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02083381 CAS-Nummer: 86847-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...