Ethyl-6-chlorimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: Apollo Scientific
Achtung

OR9842-1G OR9842-5G OR9842-10G
APOSOR9842-1GEA 21.9 EUR
APOSOR9842-1G APOSOR9842-5G APOSOR9842-10G
Ethyl-6-chlorimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-6-chlorimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₀H₉ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02186220
CAS-Nummer: 67625-38-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR