Ethyl-4-morpholinobenzenecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR9853-1G
OR9853-500MG
OR9853-5G
APOSOR9853-1GEA
168
EUR
APOSOR9853-1G
APOSOR9853-500MG
APOSOR9853-5G
Ethyl-4-morpholinobenzenecarboxylate
Ethyl-4-morpholinobenzenecarboxylate
Formel:
C₁₃H₁₇NO₃ Schmelzpunkt: 86…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06204958 CAS-Nummer: 19614-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...