Ethyl-4-morpholinobenzenecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4-(morpholin-4-yl)benzoate, Ethyl 4-Morpholinobenzoate

OR9853-1G OR9853-500MG OR9853-5G
APOSOR9853-1GEA 168 EUR
APOSOR9853-1G APOSOR9853-500MG APOSOR9853-5G
Ethyl-4-morpholinobenzenecarboxylate
Ethyl-4-morpholinobenzenecarboxylate
Formel: C₁₃H₁₇NO₃
Molecular Weight: 235.29 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06204958
CAS-Nummer: 19614-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR