tert-Butyl 4-[4-(methoxycarbonyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-[4-(methoxycarbonyl)phenyl]piperazine-1-carboxylate
OR9873-250MG
APOSOR9873-250MGEA
34.8
EUR
APOSOR9873-250MG
tert-Butyl 4-[4-(methoxycarbonyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
tert-Butyl 4-[4-(methoxycarbonyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Formel:
C₁₇H₂₄N₂O₄ Molecular Weight: 320,383 g/mol Schmelzpunkt: 165…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200865 CAS-Nummer: 158985-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...