Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate

OR9878-1G
APOSOR9878-1GEA 31.9 EUR
APOSOR9878-1G
Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate
Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate
Formel: C₈H₁₂N₂O₂
Molecular Weight: 168,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159638
CAS-Nummer: 35691-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR