N'-Hydroxyimidazo[1,2-a]pyridine-6-carboximidamide 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR9886-1G
OR9886-5G
APOSOR9886-1GEA
87
EUR
APOSOR9886-1G
APOSOR9886-5G
N'-Hydroxyimidazo[1,2-a]pyridine-6-carboximidamide 95%
N'-Hydroxyimidazo[1,2-a]pyridine-6-carboximidamide
Formel:
C₈H₈N₄O Schmelzpunkt: 191…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366550 CAS-Nummer: 885950-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...