Imidazo[1,2-a]pyridin-8-ol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR9887-500MG
APOSOR9887-500MGEA
143
EUR
APOSOR9887-500MG
Imidazo[1,2-a]pyridin-8-ol 95+%
Imidazo[1,2-a]pyridin-8-ol
Formel:
C₇H₆N₂O Schmelzpunkt: 154…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06496222 CAS-Nummer: 69214-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...