3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-((Dimethylamino)methyl)-4-hydroxy-5-methoxybenzaldehyde

OR9894-1G
APOSOR9894-1GEA 21.9 EUR
APOSOR9894-1G
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde 95%
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde
Formel: C₁₁H₁₅NO₃
Molecular Weight: 209,25 g/mol
Schmelzpunkt: 140…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03011572
CAS-Nummer: 87743-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR