L-(+)-Cystathionine

Lieferant: Apollo Scientific
Achtung

Synonyme: S-(+)-[(2R)-2-Amino-2-carboxyethyl]-L-homocysteine, (2S)-(+)-2-Amino-4-{[(2R)-2-amino-2-carboxyethyl]sulphanyl}butanoic acid

OR9895T-10MG OR9895T-50MG
APOSOR9895T-10MGEA 219 EUR
APOSOR9895T-10MG APOSOR9895T-50MG
L-(+)-Cystathionine
L-(+)-Cystathionine

Intermediate in trans-sulfuration whereby the sulfur of methionine is transferred via homocysteine to cysteine. Collection Czechoslov.

Formel: C₇H₁₄N₂O₄S
Molecular Weight: 222.262 g/mol
Schmelzpunkt: 312 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00036685
CAS-Nummer: 56-88-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR