2-(Chloromethyl)[1,3]oxazolo[4,5-b]pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR9897-1G
APOSOR9897-1GEA
132
EUR
APOSOR9897-1G
2-(Chloromethyl)[1,3]oxazolo[4,5-b]pyridine 95%
2-(Chloromethyl)[1,3]oxazolo[4,5-b]pyridine
Formel:
C₇H₅ClN₂O Schmelzpunkt: 113…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659615 CAS-Nummer: 110704-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...