2,2'-Dibromacetophenon 90%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9900-1G
OR9900-25G
OR9900-5G
APOSOR9900-1GEA
43.5
EUR
APOSOR9900-1G
APOSOR9900-25G
APOSOR9900-5G
2,2'-Dibromacetophenon 90%
2,2'-Dibromacetophenon
Formel:
C₈H₆Br₂O Siedepunkt: 90…92 °C/0,02mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD00044660 CAS-Nummer: 49851-55-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...