ω,4-Dibromacetophenon 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Brom-1-(4-bromphenyl)-1-ethanon, Tau Protein Kinase I Inhibitor, 2,4'-Dibromacetophenon, Glycogen Synthase Kinase-3β Inhibitor VII, GSK-3β Inhibitor VII, 4-Bromphenacylbromid, TPK I Inhibitor, α,4-Dibromacetophenon

OR9901-100G OR9901-25G
APOSOR9901-100GEA 133 EUR
APOSOR9901-100G APOSOR9901-25G
ω,4-Dibromacetophenon 98%
ω,4-Dibromacetophenon
Formel: C₈H₆Br₂O
Molecular Weight: 277.94 g/mol
Siedepunkt: 310
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000200
CAS-Nummer: 99-73-0
UN: 3261
ADR: 8 II
Merck Index: 13,01412

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR