3-Amino-4-chlor-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-1H-indazol-3-amine
OR9905-25G
OR9905-5G
APOSOR9905-25GEA
290
EUR
APOSOR9905-25G
APOSOR9905-5G
3-Amino-4-chlor-1H-indazol
3-Amino-4-chloro-1H-indazole
Formel:
C₇H₆ClN₃ Schmelzpunkt: 159…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00663139 CAS-Nummer: 20925-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...