tert-Butyl-4-(4-nitrobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate 95+%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-(4-nitrobenzoyl)piperazine-1-carboxylate

OR9918-1G
APOSOR9918-1GEA 223 EUR
APOSOR9918-1G
tert-Butyl-4-(4-nitrobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate 95+%
tert-Butyl-4-(4-nitrobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate
Formel: C₁₆H₂₁N₃O₅
Molecular Weight: 335,36 g/mol
Schmelzpunkt: 144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659593
CAS-Nummer: 509073-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR