Ethyl-5-formyl-2,4-dimethyl-1H-pyrrol-3-carboxylat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, Ethyl 5-Formyl-2,4-dimethyl-3-pyrrolecarboxylate, Ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate, 5-Formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester

OR9928-1G
APOSOR9928-1GEA 21.9 EUR
APOSOR9928-1G
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrol-3-carboxylat 98%
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195.22 g/mol
Schmelzpunkt: 165…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00030352
CAS-Nummer: 2199-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR