1-(4-Bromphenyl)-2-pyrrolidinon
Lieferant: Apollo Scientific
|
Achtung
|
OR9934-1G
OR9934-5G
OR9934-250MG
APOSOR9934-1GEA
0
EUR
APOSOR9934-1G
APOSOR9934-5G
APOSOR9934-250MG
1-(4-Bromphenyl)-2-pyrrolidinon
1-(4-Bromphenyl)-2-pyrrolidinon
Formel:
C₁₀H₁₀BrNO Schmelzpunkt: 98…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00138502 CAS-Nummer: 7661-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...