Ethyl-3,5-dimethyl-1H-pyrrol-2-carboxylat 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR9936-100G
OR9936-25G
APOSOR9936-100GEA
511
EUR
APOSOR9936-100G
APOSOR9936-25G
Ethyl-3,5-dimethyl-1H-pyrrol-2-carboxylat 97%
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
Formel:
C₉H₁₃NO₂ Schmelzpunkt: 125…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010645 CAS-Nummer: 2199-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...