6-Methoxy-3,4-dihydro-1H-quinolin-2-one 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methoxy-3,4-dihydroquinolin-2(1H)-one

OR9941-1G OR9941-5G OR9941-500MG
APOSOR9941-1GEA 29 EUR
APOSOR9941-1G APOSOR9941-5G APOSOR9941-500MG
6-Methoxy-3,4-dihydro-1H-quinolin-2-one 98%
6-Methoxy-3,4-dihydro-1H-quinolin-2-one
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179395
CAS-Nummer: 54197-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR