6-Methoxy-3,4-dihydro-1H-quinolin-2-one 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Methoxy-3,4-dihydroquinolin-2(1H)-one
OR9941-1G
OR9941-5G
OR9941-500MG
APOSOR9941-1GEA
29
EUR
APOSOR9941-1G
APOSOR9941-5G
APOSOR9941-500MG
6-Methoxy-3,4-dihydro-1H-quinolin-2-one 98%
6-Methoxy-3,4-dihydro-1H-quinolin-2-one
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179395 CAS-Nummer: 54197-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...