2-Ethylthio-1,3,4-thiadiazole-5-thiol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Ethylthio)-1,3,4-thiadiazole-2-thiol

OR9951-1G
APOSOR9951-1GEA 58 EUR
APOSOR9951-1G
2-Ethylthio-1,3,4-thiadiazole-5-thiol
2-Ethylthio-1,3,4-thiadiazole-5-thiol
Formel: C₄H₆N₂S₃
Molecular Weight: 178,298 g/mol
Schmelzpunkt: 120…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00186131
CAS-Nummer: 37147-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR