N-(4-Oxochroman-6-yl)acetamide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(3,4-Dihydro-4-oxo-2H-chromen-6-yl)acetamide, 6-Acetamidochroman-4-one

OR9953-1G
APOSOR9953-1GEA 292 EUR
APOSOR9953-1G
N-(4-Oxochroman-6-yl)acetamide 98%
N-(4-Oxochroman-6-yl)acetamide
Formel: C₁₁H₁₁NO₃
Molecular Weight: 205,21 g/mol
Schmelzpunkt: 164…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06202343
CAS-Nummer: 103646-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR