3-Nitro-L-tyrosin 98%

Lieferant: Apollo Scientific

Synonyme: H-Tyr(3-NO2)-OH, 3-Nitro-L-tyrosine

OR9954-25G
APOSOR9954-25GEA 78.3 EUR
APOSOR9954-25G
3-Nitro-L-tyrosin 98%
3-Nitro-L-tyrosin
Formel: C₉H₁₀N₂O₅
Molecular Weight: 226.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007123
CAS-Nummer: 621-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR