4-Iodo-6-methyl-2-methylthiopyrimidine

Lieferant: Apollo Scientific
Achtung

OR9967-250MG
APOSOR9967-250MGEA 43.7 EUR
APOSOR9967-250MG
4-Iodo-6-methyl-2-methylthiopyrimidine
4-Iodo-6-methyl-2-methylthiopyrimidine
Formel: C₆H₇IN₂S
Molecular Weight: 266,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269295
CAS-Nummer: 16879-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR